Round microcentrifuge floating bubble racks, Bel-Art ProCulture, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F188750400
BELAF188750400EA 33.9 SGD
BELAF188750400
Round microcentrifuge floating bubble racks, Bel-Art ProCulture, SP Bel-Art
Racks Tube Racks
Made especially for use with beakers and waterbaths, these space-saving racks are ideal for small quantities of tubes and for use in cell culture laboratories.
  • Buoyant TPX® brand polymethylpentene with tiny bubbles throughout for additional buoyancy
  • Insert tubes from the top and easily remove them by pressing the rack down on the bench
  • Withstands boiling for 30 minutes at 100 °C (212 °F)
  • Steam autoclavable at 121 °C (250 °F)
  • Leg length: 19 mm (³/₄"); thickness: 6,4 mm (¼")
Perfect for incubating microtubes in a beaker of water.