Racks for centrifuge bottles, Poxygrid®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F198560250
BELAF198560250EA 116 SGD
BELAF198560250
Racks for centrifuge bottles, Poxygrid®, SP Bel-Art
Racks Tube Racks
Epoxy-coated steel wire, autoclavable.
  • Supports conical and round bottom centrifuge tubes
  • The bottom rails hold the tip of the conical tube
Hole Ø: 60 mm