Disposable pestles for 1,5 ml microtubes, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F199230001
BELAF199230001EA 194 SGD
BELAF199230001
Disposable pestles for 1,5 ml microtubes, SP Bel-Art
Mortars and Pestles Pestles
These disposable plastic (PP) pestles are excellent for re-suspending bacterial cells, precipitates of nucleic acids or proteins and pellets of other materials.
  • With 1,5 ml microcentrifuge tubes of the same shape for optimal grinding
  • Pestles also available without tubes