Spill containment tray, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F246750050
BELAF246750050EA 236 SGD
BELAF246750050
Spill containment tray, SP Bel-Art
Trays/Dishes/Pots Spillage Trays/Pans
Made of Polystyrene, translucent.
  • With external flange
  • Durable
  • Liners must be ordered seperately
Protect laboratory benches. To absorb spilled liquids.
Order Now
SPECIFICATIONS
Environmentally Preferable