Glove box holders, Poxygrid®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F247390003
BELAF247390003EA 172 SGD
BELAF247390003
Glove box holders, Poxygrid®, SP Bel-Art
Glove Dispensers and Glove Box Holders
Epoxy-coated, welded steel wire racks for one, two or three boxes of disposable gloves, wipes or tissues.
  • Glove box drops into an individual retaining compartment and lifts out easily for replacement
  • Designed for mounting to a vertical surface
  • Durable, blue epoxy coated racks are steam autoclavable at 121 °C
Order Now
SPECIFICATIONS
Environmentally Preferable