Beakers, graduated, tall form, PMP (TPX) , SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F289820000
BELAF289820000EA 33.8 SGD
BELAF289820000
Beakers, graduated, tall form, PMP (TPX) , SP Bel-Art
Beakers
Crystal clear Polymethylpentene (TPX®).
  • Chemical resistant and shatterproof
  • Metric gradations molded on outside wall
  • Heat resistant to 175 °C (347 °F); steam autoclavable
Gracefully proportioned, these pitchers are ideal for general laboratory use and offer well designed spouts and large handles helpful for use with wet or dry ingredients.