Beakers, graduated, tall form, PP, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F289950000
BELAF289950000EA 325 SGD
BELAF289950000
Beakers, graduated, tall form, PP, SP Bel-Art
Beakers
Made of PP, translucent.
  • Use with wet or dry ingredients
  • Chemical resistant and shatterproof
  • Metric gradations molded on outside wall
  • Translucent polypropylene construction
  • Heat resistant to 121ºC (250ºF); steam autoclavable
Available in a large selection of sizes, these pitchers are ideal for general laboratory use and offer well designed spouts and large open handles for easy stacking.