Mercury collectors, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F362500000
BELAF362500000EA 89.5 SGD
BELAF362500000
Mercury collectors, SP Bel-Art
Spill Control Materials
The mercury globules are absorbed by the foam pad.
  • Deals safely and effectively with mercury spillages
  • Safe storage in airtight container
  • Mesh inside squeezes the mercury out of the foam, so it is immediately ready for use again