Magnetic stirring bars, PTFE, cross-shaped, Spinplus®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F371440100
BELAF371440100EA 28.1 SGD
BELAF371440100
Magnetic stirring bars, PTFE, cross-shaped, Spinplus®, SP Bel-Art
Stirrers Magnetic Stirring Bars
Spinplus® magnetic stirring bars improve speed and efficiency of mixing.
  • The '+' shape creates a deep vortex and provides stable, quiet operation
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
Certifications: Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification