Magnetic stirring bars, PTFE, octagonal, assorted, Spinpak®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F371580010
BELAF371580010EA 216 SGD
BELAF371580010
Magnetic stirring bars, PTFE, octagonal, assorted, Spinpak®, SP Bel-Art
Stirrers Magnetic Stirring Bars
Eight-sided Spinbar® octagonal magnetic stirring bars provide greater surface area and added turbulence when compared to smooth cylindrical bars.
  • Integral pivot ring on each stirring bar aids in reducing friction and chattering
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating, durable and inert
Spinbar® Restrainer fits comfortably in one hand and retains Spinbar® magnetic stirring bars up to 80 mm (3") long by holding them against vessel walls; eliminates splashing hazards when pouring.
Certifications: SGS certified.
More Product Information