Magnetic stirring bar retriever, HDPE, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F377750000
BELAF377750000EA 67.7 SGD
BELAF377750000
Magnetic stirring bar retriever, HDPE, SP Bel-Art
Stirrers Stirring Tools
This retriever is able to remove a magnetic stirring bar without coming into contact with the contents of the vessel.
  • Eliminates contamination of contents
  • Does not need to be cleaned or autoclaved after use
  • Long ribbed handle provides a secure grip