Magnetic stirring bar restrainer, Spinbar®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F377861000
BELAF377861000EA 44 SGD
BELAF377861000
Magnetic stirring bar restrainer, Spinbar®, SP Bel-Art
Stirrers Stirring Tools
The Spinbar® restrainer fits ergonomically in one hand, and when placed alongside the outer wall of a mixing vessel it retains and secures the stirring bar in position.
  • Ensures no direct contact with the stirring magnet
  • Retains magnetic stirring bars up to 80 mm long