Pipette controller, Pipette Pump™ II, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F379111025
BELAF379111025EA 79.6 SGD
BELAF379111025
Pipette controller, Pipette Pump™ II, SP Bel-Art
Pipettes Pipette Controllers
Pipette filler for glass and plastic pipettes up to 2, 10 or 25 ml.
  • Pipettes are easily inserted into a unique collar with stabilising fingers to hold pipette securely
  • Easily disassembled for cleaning
  • Colour-coded
Designed for fast and efficient single-handed operation with glass or plastic pipettes.