Mini tongs, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F379440000
BELAF379440000EA 158 SGD
BELAF379440000
Mini tongs, Sterileware®, SP Bel-Art
Tongs
PS, white. For handling food, biological, chemical or pharmaceutical samples.
  • Individually packaged, lot traceable and sterilized sample handling tongs
  • Molded from smooth white polystyrene
  • Serrated jaws securely hold small quantities of moist or dry material
  • Disposable, designed for one time use to avoid cross contamination
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904