Gas washing bottle with Fritware® dispersion disc, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H110300000
BELAH110300000EA 167 SGD
BELAH110300000
Gas washing bottle with Fritware® dispersion disc, SP Bel-Art
Bottles Gas Washing Bottles
Polypropylene gas washing bottle with fritware Polyethylene dispersion disc.
  • Gas is dispersed through a removable fitting that has a coarse grade porosity, polyethylene dispersion disc
  • Silicone stopper holds 6,3 mm (¼") O.D. tubes for entrance and exit of gas
  • 0,5 psi maximum pressure
Graduated 250 ml cylinder with a stable extended base for use in precipitating elements from gas.
Order Now
SPECIFICATIONS
Environmentally Preferable