Carboys, book bottle style, with spigot, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H118620005
BELAH118620005EA 488 SGD
BELAH118620005
Carboys, book bottle style, with spigot, SP Bel-Art
Bottles Carboys
PP, translucent.
  • Good chemical resistance
  • Good temperature resistance
This book bottle style container has a gasketed spigot threaded into the bottle. It requires only a small turn from closed position to full open and has a 9,5 mm int.Ø.opening that extends from the front of the bottle by 58 mm. The bottle has a 43 mm int.Ø neck and features a leakproof seal plug as well as a screw cap. The front is graduated in liters.
Note: Spigot has a rubber component that is incompatible with acetone and organic solvents.
Order Now
SPECIFICATIONS
Environmentally Preferable