Funnels, utility type
Supplier: Bel-Art Products, a Part of SP
H147820000
BELAH147820000EA 221 SGD
BELAH147820000
Funnels, utility type
Funnels Filtration Funnels Filter Funnels
These externally ribbed utility type funnels are made of PP.
  • Steam autoclavable at 121 °C
  • Useful for a variety of lab applications
Stem Int.Ø: 7 mm