Vibration spatulas, Quaver®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H367140000
BELAH367140000EA 190 SGD
BELAH367140000
Vibration spatulas, Quaver®, SP Bel-Art
Spatulas
Stainless steel or stainless steel with FEP coating. These spatulas are ideal for accurate additions of precious materials. Rotating the toothed thumb wheel vibrates the curved tip spatula, giving control over transfer from spatula to weighing dish.
  • Reduce transfer losses
  • Non motorised
  • Available with a plain stainless steel blade or with a brown Fluo-Kem® Teflon® FEP coating
  • Coating complies with FDA regulations governing direct food contact applications