Vibration spatulas, micro-blade, Quaverette®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H367180000
BELAH367180000EA 175 SGD
BELAH367180000
Vibration spatulas, micro-blade, Quaverette®, SP Bel-Art
Spatulas
Stainless steel or stainless steel with Teflon® FEP coating. The micro V-shaped narrow blade can be moved easily inside the small mouth containers and tight corners. Rotating the toothed thumb wheel vibrates the curved tip spatula, giving control over transfer from spatula to weighing dish.
  • Reduces transfer losses
  • Make accurate additions of precious materials as needed
Certifications: Coating complies with FDA regulations governing direct food contact applications.