Sampling scoops, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H369060000
BELAH369060000EA 658 SGD
BELAH369060000
Sampling scoops, Sterileware®, SP Bel-Art
Scoops
PS, white. Designed especially to take samples from wide mouth containers.
  • One-piece construction
  • For powder, granulates and liquids
  • Made from FDA grade materials
  • Small sizes (60 and 125 ml) have a long handle that increases the distance between the hand and sample
  • When placed on a flat surface they remain upright; can be used as weighing boats
  • Individually sterilised and packed in a polyethylene bag
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904