Sampling spatula, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H369300000
BELAH369300000EA 471 SGD
BELAH369300000
Sampling spatula, Sterileware®, SP Bel-Art
Spatulas
One-piece spatulas, moulded from FDA grade, high impact PS.
  • Sterilised by gamma irradiation, lot stamped for accurate lot tracing, shipped with a Certificate of Processing for Sterilisation
V-shaped spatula for sampling dislodge caked or crystallised reagents from deep within tall bottles or vessels having a neck diameter as small as 18 mm (³/₄"). The sturdy point is perfect for taking sterile samples during process quality inspections, and for piercing plastic-lined paper bags or similar packages.