Magnetic stirrers, battery powered, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H370170000
BELAH370170000EA 1440 SGD
BELAH370170000
Magnetic stirrers, battery powered, SP Bel-Art
Stirrers Magnetic Stirrers
This portable magnetic stirrer is made of PP.
  • Durable and chemical resistant
  • Two D batteries provide 16 days of continuous operation
Ideal for use inside incubators and when electricity is not available.
Delivery information: Supplied with Spinbar® Octagon Stirbar (BELAF371100138).