Forceps, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H379370000
BELAH379370000EA 141 SGD
BELAH379370000
Forceps, SP Bel-Art
Forceps
Stainless steel, Teflon® FEP coated (partly of fully). The brown Teflon® FEP coating on these stainless steel forceps will not scratch or mar delicate surfaces, while its anti-stick qualities prevent material build-up.
  • Coating complies with FDA regulations for direct contact with food
  • All forceps are 98 mm (3⅞") long with medium points and serrated tips