Scriber pen, Glascribe®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H441500000
BELAH441500000EA 123 SGD
BELAH441500000
Scriber pen, Glascribe®, SP Bel-Art
Pens Stylus and Scribers
The Glascribe® Pen, with retractable tungsten carbide tip, lets you inscribe on glass, ceramics, and plastic.
  • With pocket clip, so it is always handy when needed