Mounting plate for pipette canisters
Supplier: BELLCO GLASS
1288-11111
BELC1288-11111EA 0 SGD
BELC1288-11111
Mounting plate for pipette canisters
Mounting Plates
Mounting plate for pipette canisters, made from stainless steel.
  • Stabilises pipette canisters during use
  • Can be used with 38 mm Ext.Ø glass pipette canisters