Disposable caps for culture tubes and flasks, KAP-UTS™
Supplier: BELLCO GLASS
2007-13001 2007-16001 2007-16002 2007-16004 2007-16005 2007-13002 2007-13005 2007-13003 2007-13004 2007-13006 2007-16003 2007-16006
BELC2007-13001EA 0 SGD
BELC2007-13001 BELC2007-16001 BELC2007-16002 BELC2007-16004 BELC2007-16005 BELC2007-13002 BELC2007-13005 BELC2007-13003 BELC2007-13004 BELC2007-13006 BELC2007-16003 BELC2007-16006
Disposable caps for culture tubes and flasks, KAP-UTS™
Closures Test/Sample Tube Closures
Made of nontoxic polypropylene.
  • Autoclavable to 140 ºC
  • Same evaporation and sterility characteristics as stainless steel closures
  • Molded pressure fins for firm grip
Designed for culture tubes and flasks with DeLong® style necks and access ports.
Order Now
SPECIFICATIONS
Environmentally Preferable