Culture tubes, rimless
Supplier: BELLCO GLASS
2011-18150
BELC2011-18150EA 664 SGD
BELC2011-18150
Culture tubes, rimless
Tubes Culture Tubes
Manufactured from type 1 class A borosilicate glass with plain top.
  • Very good chemical resistance
  • Very good temperature resistance
Order Now
SPECIFICATIONS
Environmentally Preferable