Culture shake flasks
Supplier: BELLCO GLASS
2541-02000
BELC2541-02000EA 2190 SGD
BELC2541-02000
Culture shake flasks
Flasks Culture Flasks
Flasks, made of type 1 class A borosilicate glass, with rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation.
  • With break resistant lip to resist chipping
  • Very good chemical resistance
  • Very good temperature resistance
Designed for use with orbital shakers.