Semi automated Clinical Chemistry Analyser BeneSphera™ C-61
Supplier: BENESPHERA
20001751
BENP20001751EA 2890 SGD
BENP20001751
Semi automated Clinical Chemistry Analyser BeneSphera™ C-61
Clinical Diagnostic Supplies Clinical Analysers
BeneSphera™ C-61 semi-automated clinical chemistry analyser offers high precision and excellent reproducibility for reliable reporting.
  • Excellent QC function with the option of 2 levels of programmable controls per test
  • Double aspiration air gap after each sample prevents cross-contamination
  • Large LCD display allows for easy keypad operation
  • Memory for 60 programs and 2000 test results - suitable for running profiles