Mop head frame, small, flat, BCR® Mop 3
Supplier: BERKSHIRE
BCR.MOP3.1
BERKBCR.MOP3.1EA 534 SGD
BERKBCR.MOP3.1
Mop head frame, small, flat, BCR® Mop 3
Mops
Berkshire’s flat mop head is light and easy to maneuver, even with wet wipers attached. The stainless steel springs or plastic eyes hold multiple mop wipers.
  • Clean, light, and durable
  • High quality anodized aluminum
  • Autoclavable (plastic eyes are not autoclavable)
  • Plastic eyes for wiper attachment
Efficient cleaning of floors, walls, and other large surfaces is critical to the overall cleanliness of the production environment. BCR® mop systems replicate the most effective way to clean a surface wiping. Berkshire’s mop systems are easy to use, highlyMore Product Information