Glass stoppers for iodine flasks, Quickfit®
Supplier: SciLabware
SIB24
BIBBSIB24EA 259 SGD
BIBBSIB24
Glass stoppers for iodine flasks, Quickfit®
Stoppers Glass Stoppers
Quickfit® iodine flask stoppers.
  • Extended handle which projects well above the liquid seal
  • Available with either 24/29 or 29/32 ground cone joints
For use with Quickfit® Iodine flasks.