Pipette tips, gel loading
Supplier: BIOTIX
2016 2016.S
BITX2016EA 1200 SGD
BITX2016 BITX2016.S
Pipette tips, gel loading
Pipette Tips
These pipette tips are specially designed to help load the thinnest sequencing gels.
  • Extended length
  • Universal fit: Ideal for single & multi-channel pipettors
  • Eco-friendly racks