Mini tube system
Supplier: BIOTIX
2600.8.X 2601.8.X 2601.8S.X 2601.S.X 2601.X
BITX2600.8.XEA 65.3 SGD
BITX2600.8.X BITX2601.8.X BITX2601.8S.X BITX2601.S.X BITX2601.X
Mini tube system
Tubes Microtubes
PP.
This minitube system is ideal for microplate-to-tube transfer or manual-to-robotic-process transfer. 1,1 ml strip minitubes come pre-racked in a standard 96-well format (8×12) for convenient, multi-channel pipetting. Alphanumerically labeled, non-reversing rack and lid ensure positive sample identification. Condensation rings above each tube minimize cross-contamination between wells. System is completely autoclavable (except for optional mini-cap strips). The rack base is fully compatible with all leading robotic instruments.