Strips of 8 PCR tubes with detached cap strips
Supplier: BIOTIX
3426.8.X
BITX3426.8.XEA 162 SGD
BITX3426.8.X
Strips of 8 PCR tubes with detached cap strips
Tubes PCR Tubes
Made of virgin polypropylene copolymer, clear.
  • Sterile or non sterile
  • Autoclavable
  • Excellent chemical resistance
Specifically designed for PCR, these tubes feature uniform microthin walls for efficient heat transfer and fast cycling times. Tubes and caps are compatible with most leading thermal cyclers.
Order Now
SPECIFICATIONS
Environmentally Preferable