Pipette and filter tips for Rainin® LTS® pipettes, xTIP4™, low retention
Supplier: BIOTIX
63300008
BITX63300008EA 113 SGD
BITX63300008
Pipette and filter tips for Rainin® LTS® pipettes, xTIP4™, low retention
Pipette Tips
xTIP4™ is the only Rainin® LTS® compatible tip engineered with StarStop, a patented positive stop feature. This feature minimizes ejection force and reduces the risk of repetitive stress injuries (RSI).
  • FlexFit sealing technology lowers the forces necessary to attach and eject tips with a secure seal
  • All tips are Low binding to maximize sample recovery and increase accuracy
  • Blade tip reduces the need for tip touch off and increases accuracy and precision
  • Compatible with both multi- and single channel Rainin® LTS® pipettes
  • Filter tips are made from
More Product Information