Robotic tips for Agilent automated platforms
Supplier: BIOTIX
63300094 63300093
BITX63300094EA 0 SGD
BITX63300094 BITX63300093
Robotic tips for Agilent automated platforms
Pipette Tips
Biotix robotic tips for Agilent™ VPrep® and Bravo® platforms are engineered to ensure complete compatibility, requiring no changes in programming scripts in the platform's liquid handling software suite.
  • Made of medical-grade virgin PP
  • Tips are BioReady™ certified to be free of RNase, DNase, and Endotoxins (pyrogens)
  • Available as standard or filtered tips
Manufactured to the highest engineering standards for robotic systems, the ultra-straight tips target the centre of microplate wells precisely, delivering exceptional accuracy and precision without risk of jamming.