Pipette tips
Supplier: BIOTIX
63302158 63302160 2107.Y 63302165 2102.Y 63302163 63300532 63300566
BITX63302158EA 89.9 SGD
BITX63302158 BITX63302160 BITX2107.Y BITX63302165 BITX2102.Y BITX63302163 BITX63300532 BITX63300566
Pipette tips
Pipette Tips
These pipette tips are made from virgin PP or exclusive S3 polymer.
  • Polymer tips retain virtually no fluids, allowing the user to dispense truly accurate sample volumes
  • Environmentally friendly packaging: some tips available in ESP reload systems
  • Tips are certified free of RNase, DNase, DNA and endotoxins
  • Graduated sides
The exclusive S3 polymer delivers to within 0,1% of indicated volume, eliminating sample hold-up and ensuring a more precise application. This proprietary technology is essential for quantitative assays where sample retention canMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Safer Human and Environmental Health,Sustainable Packaging