Chlorobenzene ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™

Supplier: BURDICK JACKSON
Warning

043-4
BJAA043-4EA 2890 SGD
BJAA043-4
Chlorobenzene ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Chlorobenzene

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: C₆H₅Cl
Boiling Pt: 132 °C
Melting Pt: –46 °C
Density: 1,1058 g/cm³ (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.5242 - 1.5256
UV Absorbance @ 300 nm 0.05 AU max.
UV Absorbance @ 325 nm 0.04 AU max.
UV Absorbance @ 350 nm 0.02 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 287 nm max.
Water by Karl Fischer Titration 0.03 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR