Isopropyl myristate, B&J Brand™ for sterility testing of ophthalmic ointments, Burdick & Jackson™

Supplier: BURDICK JACKSON

Synonyms: Tetradecanoic acid isopropyl ester, Myristic acid isopropyl ester, Isopropyl Myristate, Isopropyl tetradecanoate, Tetradecylic acid isopropyl ester

225-4
BJAA225-4EA 1630 SGD
BJAA225-4
Isopropyl myristate, B&J Brand™ for sterility testing of ophthalmic ointments, Burdick & Jackson™
Isopropyl myristate

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 193 °C (26.7 hPa)
Melting Pt: –3 °C
Density: 0.853 g/cm³ (20 °C)
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00008982
CAS Number: 110-27-0
EINECS: 203-751-4
Merck Index: 13,05235

Order Now

Specification Test Results

Appearance Passes test
Identification by IR Passes test
pH of Water Extract 6.5 min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR