Methanol ≥99.9%, B&J Brand™ for purge and trap GC analysis, for determination volatile organic compounds, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

232-235
BJAA232-235EA 1790 SGD
BJAA232-235
Methanol ≥99.9%, B&J Brand™ for purge and trap GC analysis, for determination volatile organic compounds, Burdick & Jackson™
Methanol

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: CH₃OH
Boiling Pt: 64,7 °C
Melting Pt: –98 °C
Density: 0,792 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.3280 - 1.3288
Residue 1 mg/L max.
Volatile Organic Compounds Passes test
Water by Karl Fischer Titration 0.3 % max.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR