Acetonitrile : Water (50:50)% (v/v), LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

435-4
BJAA435-4EA 4170 SGD
BJAA435-4
Acetonitrile : Water (50:50)% (v/v), LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Acetonitrile : Water

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 82 °C (solvent)
Density: 0,7857 g/cm³ (20 °C) (pure substance)
Flash Pt: 3 °C
Storage Temperature: Ambient
EINECS: 200-835-2
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Water (v/v) 49.0 - 51.0%
Acetonitrile (v/v) 49.0 - 51.0%
UV Cutoff 186 nm
Residue < 1 mg/L
Color (APHA) < 10
HPLC Gradient @ 220 nm < 0.005 AU
HPLC Gradient @ 254 nm < 0.001 AU


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR