Reagent alcohol, anhydrous ACS, meets analytical specification of USP for HPLC, for spectrophotometry (denatured with methanol and isopropanol), Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

Synonyms: Reagent alcohol

AH090-4
BJAAAH090-4EA 1810 SGD
BJAAAH090-4
Reagent alcohol, anhydrous ACS, meets analytical specification of USP for HPLC, for spectrophotometry (denatured with methanol and isopropanol), Burdick & Jackson™
Alcohol

Specially Denatured Alcohol Formula 3-A-95parts by vol., and Isopropyl Alcohol-5 parts by vol.


  • Total alcohol: ~100%
  • Ethyl alcohol: 89-92%
  • Denaturant: Methyl alcohol and 4-6% 2-propanol

Boiling Pt: 78,32 °C (main component)
Melting Pt: –114,1 °C (main component)
Density: 0,7890 g/cm³ (20 °C)
Flash Pt: 15 °C (main component)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Color (APHA) 10 max.
GC Assay (Ethanol + Methanol) 94 - 96 %
GC Assay (Isopropyl Alcohol) 4 - 6 %
Residue 1 mg/L max.
Specific Gravity (25ºC/25ºC) 0.785 - 0.795
UV Cutoff 210 nm max.
Water by Karl Fischer Titration 0.2 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR