Ethyl acetate ≥99.5% ACS, meets analytical specification of USP for HPLC, for spectrophotometry

Supplier: BURDICK JACKSON
Danger

Synonyms: Acetic acid ethyl ester

AH100-4
BJAAAH100-4EA 1510 SGD
BJAAAH100-4
Ethyl acetate ≥99.5% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Ethyl acetate

Meets USP Reagent Specifications


  • ACS certified
  • Lot-to-lot consistency
  • HPLC grade controlled for UV absorbance, GC assay, water content and non-volatile residue

Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 11.5 %(V)
Melting Pt: –84 °C
Density: 0.9006 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10 max.
GC Analysis 99.5 % min.
Peroxides (as H₂O₂) at time of packaging 2 mg/L max.
Refractive Index (20ºC) 1.3720 - 1.3726
Residue 3 mg/L max.
Substances Darkened by Sulfuric Acid Passes test
Titrable Acid 0.9 µeq/g max.
UV Cutoff 256 nm max.
Water by Karl Fischer Titration 0.03 % max.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR