Hexane (mixture of isomers) ≥99.9% (n-Hexane > 60%) ACS, meets analytical specification of USP for HPLC, for spectrophotometry

Supplier: BURDICK JACKSON
Danger

Synonyms: Hexanes, Isohexane

AH212-4
BJAAAH212-4EA 1480 SGD
BJAAAH212-4
Hexane (mixture of isomers) ≥99.9% (n-Hexane > 60%) ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Hexane (mixture of isomers)

  • ACS certified
  • Lot-to-lot consistency
  • HPLC grade controlled for UV absorbance, GC assay, water content and non-volatile residue

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 7,7 %(V)
Melting Pt: –95 °C
Density: 0,659…0.673 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10 max.
Water-Soluble Titrable Acid 0.3 µeq/g max.
GC Analysis (n-Hexane) 60 % min.
GC Analysis (total C6 isomers) 99.9 % min.
Residue 3 mg/L max.
Sulfur Compounds (as S) 0.005 % max.
Thiophene Passes test
UV Absorbance @ 225 nm 0.05 AU max.
UV Cutoff 195 nm max.
Water by Karl Fisher Titration 0.01 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR