Water for HPLC, for spectrophotometry

Supplier: BURDICK JACKSON

NPAH365-20 AH365-4
BJAANPAH365-20EA 988 SGD
BJAANPAH365-20 BJAAAH365-4
Water for HPLC, for spectrophotometry
Water

For HPLC and spectrophotometry. UV cutoff 190nm.
Not ACS Certified


  • Lot-to-lot consistency
  • HPLC grade controlled for UV absorbance, GC assay, water content and non-volatile residue

Formula: H₂O
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0,9971 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

HPLC Gradient @ 200 nm 0.005 AU max.
UV Absorbance @ 250 nm 0.005 AU max.
Microbial Contamination Not detected
Refractive Index (20ºC) 1.3320 - 1.3340
Residue 1 mg/L max.
Specific Conductance (25ºC) 2 Micromhos max.
Substances Reducing Permanganate Passes test
Total Organic Carbon 50 ppb max.
UV Absorbance @ 190 nm 0.01 AU max.
UV Absorbance @ 200 nm 0.01 AU max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR