Dichloromethane ≥99.9% stabilised, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™

Supplier: BURDICK JACKSON
Warning


Stabilization: Stabilised with cyclohexene

BB301-4
BJAABB301-4EA 1350 SGD
BJAABB301-4
Dichloromethane ≥99.9% stabilised, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Dichloromethane

BioSyn products are ideal for DNA, RNA and oligonucleotide synthesis. All solvents and regents are filtered (0.2-micron) and tested for oxidation reage.


  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity

Formula: CH₂Cl₂
Boiling Pt: 40 °C
Melting Pt: –95 °C
Density: 1,33 g/cm³
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acidity (as HCl) 1 mg/L max.
Chloride 10 mg/L max.
GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.4236 - 1.4246
Residue 1 mg/L max.
UV Absorbance @ 240 nm 0.1 AU max.
UV Absorbance @ 250 nm 0.01 AU max.
UV Cutoff 233 nm max.
Water by Karl Fischer Titration 20 ppm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR