Hexane (mixture of isomers) ≥99.9% (n-Hexane > 95%), GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

GC215-4
BJAAGC215-4EA 2380 SGD
BJAAGC215-4
Hexane (mixture of isomers) ≥99.9% (n-Hexane > 95%), GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
Hexane (mixture of isomers)

  • Less than 1 ppb trace contamination for
  • Semi-volatiles with a molecular weight
  • Greater than toluene (92)
  • Ideal for EPA protocols
  • Actual lot chromatograms available upon request

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 7,7 %(V)
Melting Pt: –95 °C
Density: 0,659…0.673 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC (as lindane) 2 ppt max.
Flame Ionization GC (as nonane) 1 ppb max.
GC Analysis (n-Hexane) 95 % min.
GC Analysis (total C6 isomers) 99.9 % min.
Residue 1 mg/L max.
Water by Karl Fischer Titration 0.01 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR