Water, B&J Brand™ LC-MS for LC-MS, for HPLC, Burdick & Jackson™

Supplier: BURDICK JACKSON

Synonyms: Hydrogen oxide

LC365-1 LC365-2.5
BJAALC365-1EA 840 SGD
BJAALC365-1 BJAALC365-2.5
Water, B&J Brand™ LC-MS for LC-MS, for HPLC, Burdick & Jackson™
Water

HIGH PURITY SOLVENTS, LC-MS GRADE SOLVENTS


  • Extensive quality control to ensure that they meet our specifications
  • Low solvent and metal impurities for reduced interference with delicate, minute or unknown samples
  • Minimal organic and trace metal contamination, which results in higher ionization efficiency, lower adduct formation and easier data interpretation
  • LC-MS tested

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.9971 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Fluorescence (as quinine base) 0.3 ppb max.
HPLC Gradient Elution Passes test
LC-MS Negative Mode (as 4-nitrophenol) 50 ppb max.
LC-MS Positive Mode (as caffeine) 100 ppb max.
LC-MS Positive Mode (as reserpine) 50 ppb max.
Refractive Index (20ºC) 1.3325 - 1.3335
Residue 1 mg/L max.
Trace Metals Passes test
Ultraviolet Absorbance Test Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR