Hexane (mixture of isomers) ≥99.9% (n-Hexane > 60%), Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

Synonyms: Hexanes, Isohexane

PP211-19 PP211-204
BJAAPP211-19EA 1300 SGD
BJAAPP211-19 BJAAPP211-204
Hexane (mixture of isomers) ≥99.9% (n-Hexane > 60%), Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Hexane (mixture of isomers)

  • Low residue
  • Purified solvent
  • ACS General Use certified

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 7,7 %(V)
Melting Pt: –95 °C
Density: 0,659…0.673 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay by GC Analysis (n-Hexane) 60 % min.
Assay by GC Analysis (total C6 isomers) 98.5 % min.
Color (APHA) 10 max.
Residue after Evaporation 5 mg/L max.
Sulfur Compounds (as S) 0.005 % max.
Thiophene Passes test
Water by Karl Fischer Titration 0.01 % max.
Water-Soluble Titrable Acid 0.3 µeq/g max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR