Wire baskets
Supplier: BOCHEM
10050 10051 10052 10053 10054 10055 10057 10056
BOCH10050EA 162 SGD
BOCH10050 BOCH10051 BOCH10052 BOCH10053 BOCH10054 BOCH10055 BOCH10057 BOCH10056
Wire baskets
Baskets Wire Baskets
Stainless steel.
  • Spot-welded grid 8×8×1 mm
  • Reinforced edges
  • Stowable handle
These round baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
Order Now
SPECIFICATIONS
Environmentally Preferable